Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z743790

ART® Non-Filtered Low Retention Tip

ART 200 REACH, size 200 μL, hinged rack, non-sterile

Synonym(s):

aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 768 EA
$735.00

$735.00


Check Cart for Availability


Select a Size

Change View
5 X 768 EA
$735.00

About This Item

UNSPSC Code:
41121603
NACRES:
NB.02
volume:
200 μL
sterility:
non-sterile
feature:
ART 200 REACH
hinged rack
packaging:
pkg of 5X768 ea
manufacturer/tradename:
Molecular BioProducts 3542-05-HR

$735.00


Check Cart for Availability

sterility

non-sterile

feature

ART 200 REACH
hinged rack

packaging

pkg of 5X768 ea

manufacturer/tradename

Molecular BioProducts 3542-05-HR

technique(s)

PCR: suitable

size

200 μL

volume

200 μL

Looking for similar products? Visit Product Comparison Guide

General description

Supplied in environmentally friendly packaging. the non-filtered low retention reload system combines individually packaged blister tubs made of recycled plastic and new hinged racks. The push down blister tub easily inserts the ART barrier tips into the hinged racks providing stackable, space saving racks that offer easy, one-handed use. Empty hinged rack part numbers allow for easy conversion to the ART Reload System. Manufactured according to the industry′s highest quality standards, ART tips are certified as RNase, DNase and Pyrogen free, and are ideal for the pipetting of radioactive samples, nucleic amplification procedures, or any applications where critical sample handling is required. ART™ non-filtered low retention pipette tips ensure reduced sample loss and maximize sample recovery.

Features and Benefits

  • Low retention non-filter tips
  • Tip′s hydrophobic inner surface facilitates accurate data.
  • Hinged racks allow for easy access to the tips.
  • Compatible with radioactive compounds, viscous materials, PCR reagents, and more.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service