Z743701
ART® Barrier Tip
ART 1000G, size 1000 μL, individually wrapped, non-sterile
Synonym(s):
aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip
Select a Size
$206.00
Select a Size
About This Item
ART 1000G
$206.00
Recommended Products
sterility
non-sterile
feature
individually wrapped
ART 1000G
packaging
pkg of 500 ea
manufacturer/tradename
Molecular BioProducts 2079GIW
technique(s)
PCR: suitable
size
1000 μL
volume
1000 μL
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- ART barrier tips ensure complete protection against aerosols and liquid contamination.
- The tips are free from DNase, RNase, DNA, ATP, PCR inhibitors, and endotoxins.
- Stackable barrier tip packaging enables tips to pass through offset holes in the lower insert.
- Blister sleeve guide rails facilitate smooth transfer of tip inserts into the rack.
- An outer sleeve shields tips from external contamination during the refilling of inserts.
- The tip packaging option helps reduce plastic waste.
- The rigid insert design ensures reliable tip loading.
- Lot number and description are printed on every insert for easy tracking and identification.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service