Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z743294

Julabo® adaptors

twin distributing with barbed fittings, for 12 mm i.d. tubing, pack of 1

Synonym(s):

barbed adaptor

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$127.00

$127.00


Estimated to ship onJune 16, 2025FromMILWAUKEE



Select a Size

Change View
1 EA
$127.00

About This Item

UNSPSC Code:
41103706
NACRES:
NB.77

$127.00


Estimated to ship onJune 16, 2025FromMILWAUKEE


packaging

pack of 1

manufacturer/tradename

Julabo 8970471

Looking for similar products? Visit Product Comparison Guide

General description

For use with Julabo® circulators, twin distributing adapter with barbed fittings for tubing 12 mm ID, pack of 1.

Legal Information

Julabo is a registered trademark of Julabo USA, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service