Key Documents
Z743083
Julabo® stainless steel basket for 0.33/0.5 L bottles, for use with CD-1000F
pack of 1 ea
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
Product Z743083 is not currently sold in your country. Contact Technical Service
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9970337
General description
Stainless steel basket that can hold a maximum of 20 0.33/0.5 L bottles, suitable for temperatures up to 100°C.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)