Key Documents
Z742902
Whatman® HPLC column for separating taxanes of Pacific Yew trees
About This Item
UNSPSC Code:
41115709
Pricing and availability is not currently available.
packaging
pkg of 1 ea
manufacturer/tradename
Whatman 4601-1001
Whatman Article No. 28419609 (US reference)
General description
For great discoveries such as Taxol, Whatman technology optimally separates the closely eluting taxanes of Pacific yew trees.
Whatman worked closely with two leading customers to develop a specific bonded phase that achieves baseline resolution of the paclitaxel molecule from its closest impurity. Each lot of TAC 1 (Taxane Analysis Column) is tested with a paclitaxel chromatographic purity separation to ensure the best possible reproducibility.
*Richheimer SL et al. Anal Chem. 1992; 64: 2323-2326
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)