Key Documents
Z741983
Büchi® Rotavapor R-100
cold trap, with interface I-100, vacuum pump V-100, recirculating chiller F-100, AC/DC input 120 AC, US 3-pin plug
Synonym(s):
Rotavapor
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
Product Z741983 is not currently sold in your country. Contact Technical Service
joint
joint SJ24/40
packaging
set of 1
manufacturer/tradename
Büchi 11100C2012
General description
The rotary evaporator R-100 is a high-quality, entry level Rotavapor to meet the essential needs in classical laboratory applications. It is capable of a manually adjustable rotation speed of 20 to 280 rpm and equipped with an action jack lift for manual lifting of the evaporating flask. Included is the selected condenser, 1 L receiving and evaporating flasks, and a vapor duct. All accessories are included for out of the box usage.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)