Recommended Products
feature
CE compliant
battery included (4 AAA and microphone windscreen)
(WEEE Compliant)
packaging
pkg of 1 ea
manufacturer/tradename
Extech Instruments 407730
size
9 in. × 2.2 in. × 1.7 in., 230x57x44 mm
weight
5.6 oz160 g
General description
- ±2dB accuracy with 0.1dB resolution
- A&C weighting
- AC analog output
- Record Max/Min values over time
- Auto power off and Max Hold functions
- Utilizes 0.5"(12.7mm) condenser microphone
- Tripod mountable
- 40 to 130dB measuring range
- Fast/slow response time
- Tripod mountable
Applications:
- Monitoring workplace machinery
- Installing audio and alarm systems
- Product noise certification and reduction
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service