Key Documents
Z741854
Extech Video Borescope Inspection Camera
Wireless
About This Item
UNSPSC Code:
41100000
NACRES:
NB.61
feature
CE compliant
battery included (4 AA)
(WEEE Compliant)
packaging
pkg of 1 ea
manufacturer/tradename
Extech Instruments BR250
size
3.9 in. × 2.7 in. × 1 in., 100 x 70 x 25mm
weight
4.9 oz140 g
General description
Extech′s waterproof borescopes are completely submersible in water down to 1 meter for 1 hour. The built-in bright LED lamps illuminate dimly lit areas.
Applications:
  • Underwater & marine applications
  • Automotive maintenance/repair
  • Pipe inspection/repair
  • Industrial equipment inspection
  • Construction sites, Workshops, & Laboratories
  • Automotive Plumbing/Pipes - Home Inspection
Choose from one of the most recent versions:
Certificates of Analysis (COA)