Key Documents
Z741848
Extech Dual-Range AC Voltage Detector with Flashlight
About This Item
UNSPSC Code:
41100000
feature
CE compliant: no
battery included (2 AAA)
(WEEE Compliant)
packaging
pkg of 1 ea
manufacturer/tradename
Extech Instruments DV25
size
6.25 in. × 1 in. , 159x25 mm
weight
1.48 oz42 g
General description
Pocket size Non-Contact Voltage Detector with two selectable ranges
  • Non-contact detection of AC Voltage in 2 ranges:
24 to 1000VAC
100 to 1000VAC
  • For use on 50/60Hz circuits
  • Audible and Visual voltage indication
  • Built-in bright flashlight with On/Off button
  • Rugged double-molded housing
  • Tip fits into outlets or against wire insulation testing
Choose from one of the most recent versions:
Certificates of Analysis (COA)