Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z741838

Extech Portable Sucrose Brix Refractometer (0 to 32%)

with ATC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.61

description

Automatic Temperature Compensation (ATC) from 10 to 30°C

feature

(WEEE compliant)

packaging

pkg of 1 ea

manufacturer/tradename

Extech Instruments RF15

measuring range

0 (sucrose (Brix) (10 to 30°C))

size

6.6 in. × 1.6 in. × 1.2 in., 170 x 40 x 30mm

weight

7 oz200 g

Looking for similar products? Visit Product Comparison Guide

General description

  • Measure the concentration of Sugar, Salt, Lubricants, Antifreeze, Cutting Fluids & more
  • Easy to operate refractometers provide accurate and repeatable measurements on easy to read scales
  • Automatic temperature compensation models available for sugar, salt, lubricants, cutting fluids, and antifreeze
  • Requires only 2 or 3 drops of solution
  • The prism and lens with a simple focus adjustment provides repeatable results
  • Complete with case, calibration screwdriver, and calibration solution


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service