Key Documents
Z741831
Extech Datalogging Thermocouple Thermometer
4-Channel
About This Item
UNSPSC Code:
42182209
NACRES:
NB.79
feature
CE compliant
battery included (6 AA)
(WEEE Compliant)
packaging
pkg of 1 ea
manufacturer/tradename
Extech Instruments SDL200
size
7.2 in. × 2.9 in. × 1.9 in., 182 x 73 x 47.5mm
weight
17.6 oz500 g
General description
Applications:
  • Heat distribution in thermal chambers
  • Quality Control temperature monitoring
  • HVAC and Refrigeration
  • Cooling systems
  • Refrigeration
  • Core temperature in ovens
  • Automotive design
Features:
  • Stores 99 readings manually and 20M readings via 2G SD card
  • Records readings with real date and time stamp
  • User programmable sampling rate: 1 to 3600 seconds
  • Min/Max and Data Hold functions
  • Auto power off with disable function
  • Built-in PC interface
Choose from one of the most recent versions:
Certificates of Analysis (COA)