Key Documents
Z741813
Extech 4 - 1 Environmental Meter
About This Item
UNSPSC Code:
41100000
NACRES:
NB.61
feature
CE compliant
battery included
WEEE Compliant
packaging
pkg of 1 ea
manufacturer/tradename
Extech Instruments 45170
battery power
9V
General description
Pocket-size rugged meter for Humidity, Temperature, Air Velocity and Light
Features:
  • Ergonomic pocket size housing with large dual LCD simultaneous display of Temperature and Air Velocity or Relative Humidity
  • Characters on display reverse direction depending on Hygro-Thermo-Anemometer or Light Mode
  • Data Hold to freeze the displayed value
  • Records Min/Max readings
  • Built-in low friction vane wheel improves accuracy of air velocity in ft/min, MPH, m/s, km/h, and Knots
  • Built-in precision thin-film capacitance humidity sensor for fast response
  • Built in thermistor for ambient temperature measurements
High temperature measurements using Type K thermocouple
  • Utilizes precision photo diode and color correction filter
  • Cosine and color corrected light measurements
  • Auto power off, Low Battery and overrange indication
  • Complete with built-in Humidity sensor, Light sensor and Vane wheel, wrist strap, and 9V battery
Choose from one of the most recent versions:
Certificates of Analysis (COA)