Z741806
Extech Desktop Indoor Air Quality Carbon Dioxide (CO2), Temp, Humidity Monitor
Datalogger
About This Item
AC/DC input
100 - 240 V AC (with US, EU, UK, and AU plug)
feature
CE compliant
WEEE compliant
packaging
pkg of 1 ea
manufacturer/tradename
Extech Instruments CO220
humidity measuring range
0.1-99.9 %RH
measuring range
0-9999 ppm (CO2)
temperature measuring range
-10-60 °C
size
6.1 in. (155 mm) × 3.4 in. (87 mm) × 3.2 in. (81 mm)
weight
5.8 oz (165 g)
Looking for similar products? Visit Product Comparison Guide
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service