Key Documents
Z741791
Extech ExStik®
pH Meter
About This Item
UNSPSC Code:
41100000
NACRES:
NB.79
description
Waterproof
feature
CE compliant
battery included
WEEE compliant
packaging
pkg of 1 ea
manufacturer/tradename
Extech Instruments PH100
pH measuring range
0.00-14.00, accuracy: ±0.01
size
1.4 in. (35.6 mm) × 6.8 in. (172.7 mm) × 35.6 in. (40.6 mm)
weight
3.85 oz (110 g)
battery power
4 Eveready ECR2032 or equivalent
Legal Information
ExStik is a registered trademark of FLIR Commercial Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)