Key Documents
Z741117
Stuart SCT1 temperature controller
sensor type, PT100
About This Item
UNSPSC Code:
41112200
material
stainless steel probe
feature
Designed for use with the Stuart Undergrad range of stirrer hotplates
temperature measuring range
20-200 °C, accuracy: ±0.5 °C, resolution: 1%
W × D × H
90 mm × 75 mm × 123 mm
weight
0.3 kginc. probe
General description
  • Accurate liquid temperature control
  • Built in retort fitting and probe holder
  • Detachable temperature probe
  • Bright, easy to read LED display
  • Compact and light weight
  • Comprehensive range of accessories
The Stuart SCT1 temperature controller is the ideal instrument for the accurate temperature control of aqueous or oil based samples in the laboratory. Designed for use with the Stuart Undergrad range of stirrer hotplates, the SCT1 can be used either as a precise controller of temperature up to a maximum of 200°C or as a digital thermometer up to 325°C.
The SCT1 temperature controller features an in-built clamp allowing the controller to be mounted either on a horizontal or vertical retort rod, providing flexible positioning of the controller. In addition, the stainless steel temperature probe, used to measure the temperature of the sample, is detachable, allowing the main body of the SCT1 temperature controller to be positioned away from potentially damaging fumes.
The SCT1 temperature controller regulates the hotplate to accurately control the temperature of the sample, which is set via the large LED display. When not in use the temperature probe can be held securely by the in-built probe holder.
Legal Information
Stuart is a trademark of Bibby Sterlin Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)