Key Documents
Z740961
Sigma-Aldrich
Aldrich® Essentials culture tube with screw cap and rubber liner
capacity 15 mL, O.D. × L 15 mm × 150 mm
About This Item
UNSPSC Code:
41121706
NACRES:
NB.11
material
borosilicate glass tube
round bottom
screw top cap
screw top neck
transparent tube
feature
neck thread external thread
packaging
pack of 72 ea
manufacturer/tradename
Sigma-Aldrich
O.D. × L
15 mm × 150 mm
capacity
15 mL
General description
Clear borosilicate glass, round bottom culture tubes with thread top and black phenolic cap. Cap has plain rubber liner. Conforms to ASTM E982.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)