Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z740835

Sigma-Aldrich

Aldrich® Essentials graduated cylinder, class B, student grade

capacity 50 mL, subdivision, 1.0 mL, tol. 1.0 mL, Graduated "To Contain", plastic bumper and base

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121805
NACRES:
NB.43

feature

subdivision 1.0 mL

packaging

pkg of 24 ea

tol. measuring range

1.0 mL

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

General description

Clear borosilicate glass graduated cylinders with a removable plastic base and plastic protection bumper to guard against breakage. Class B accuracy. Graduated “To Contain”.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service