Key Documents
Z740699
Sigma-Aldrich
Aldrich® Essentials boiling flask, flat-bottom
capacity 250 mL, neck diam. 35 mm
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass flask
flat bottom
single-neck
packaging
pkg of 12 ea
capacity
250 mL
neck diam.
35 mm
General description
Clear borosilicate glass flat bottomed boiling flasks. Narrow long neck and narrow mouth with beaded rim. Large white marking spot.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)