Key Documents
Z740692
Sigma-Aldrich
Aldrich® Essentials boiling flask, round-bottom
capacity 500 mL, neck diam. 34 mm
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass flask
round bottom
single-neck
packaging
pkg of 6 ea
capacity
500 mL
neck diam.
34 mm
General description
Clear borosilicate glass round bottomed, (Florence), boiling flasks. Narrow long neck and narrow mouth with tooled beaded rim. Large white marking spot and capacity. Designed to meet ASTM E1403 specifications.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)