Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z740651

Sigma-Aldrich

Aldrich® Essentials Erlenmeyer flask, narrow mouth

capacity 100 mL, stopper joint: 5, volume 50-100 mL, accuracy: 25 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804

material

borosilicate glass flask
flat bottom

joint

joint: stopper5

sterility

non-sterile

feature

without cap

packaging

pkg of 48 ea

volume measuring range

50-100 mL, accuracy: 25 mL

capacity

100 mL

Looking for similar products? Visit Product Comparison Guide

General description

Clear borosilicate glass, standard wall Erlenmeyer flasks for standard laboratory use. Narrow mouth opening with beaded rim. Reference graduations and marking spot in white. ASTM E 1404, DIN 12380.

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service