Key Documents
Z740647
Sigma-Aldrich
Aldrich® Essentials Erlenmeyer flask, narrow mouth
capacity 25 mL, volume 10-25 mL, accuracy: 5 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass flask
flat bottom
joint
joint stopper0
sterility
non-sterile
feature
without cap
packaging
pkg of 48 ea
volume measuring range
10-25 mL, accuracy: 5 mL
capacity
25 mL
General description
Clear borosilicate glass, standard wall Erlenmeyer flasks for standard laboratory use. Narrow mouth opening with beaded rim. Reference graduations and marking spot in white. ASTM E 1404, DIN 12380.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)