Z740204
Amicon® Ultra-15
PLTK Ultracel-PL membrane, 30 kDa
Synonym(s):
Amicon Ultra centrifugal filter units
About This Item
Recommended Products
material
polyethylene cap
polypropylene tube
regenerated cellulose membrane
styrene-butadiene housing
sterility
non-sterile
feature
30 kDa
packaging
pack of 24 ea
manufacturer/tradename
Millipore UFC903024
tube size
~50 mL
working volume
15 mL
pore size
30 kDa MWCO
Looking for similar products? Visit Product Comparison Guide
General description
Swing bucket rotorL 4,000 x g; Fixed angle rotor (35°): 5,000 x g.
- High recovery Ultracel regenerated cellulose membrane in a range of molecular weight cut-offs.
- High retentate recovery of >90%
- Vertical membrane reduces concentration polarization for ultra-fast spin times (as fast as 10-15 mins)
- Heat-sealed membrane minimizes downstream extractables
- 100% integrity tested for reliable performance
- Convenient sample monitoring with translucent housing and volume graduations
- Direct pipettor sample access eliminates processing step to recover concentrate
- High concentration factors of 80 - 100X
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service