Key Documents
Z740178
Millipore
Amicon® Ultra-0.5
Ultracel-50 membrane, 50 kDa
About This Item
UNSPSC Code:
41104816
NACRES:
NB.24
material
regenerated cellulose membrane
styrene-butadiene housing
sterility
non-sterile
feature
50 kDa
packaging
pack of 96 ea
manufacturer/tradename
Millipore UFC505096
tube size
~1.5 mL
working volume
0.5 mL
pore size
50 kDa MWCO
Legal Information
Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)