Key Documents
Z740172
Millipore
Amicon® Ultra-0.5
Ultracel-10 membrane, 10 kDa
About This Item
UNSPSC Code:
41104816
NACRES:
NB.24
material
regenerated cellulose membrane
styrene-butadiene housing
sterility
non-sterile
feature
10 kDa
packaging
pack of 500 ea
manufacturer/tradename
Millipore UFC5010BK
tube size
~1.5 mL
working volume
0.5 mL
pore size
10 kDa MWCO
Legal Information
Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)