Key Documents
Z740169
Millipore
Amicon® Ultra-0.5
Ultracel-3 membrane, 3 kDa
About This Item
UNSPSC Code:
41104816
NACRES:
NB.24
Pricing and availability is not currently available.
material
regenerated cellulose membrane
styrene-butadiene housing
sterility
non-sterile
feature
3 kDa
packaging
pack of 96 ea
manufacturer/tradename
Millipore UFC500396
tube size
~1.5 mL
working volume
0.5 mL
pore size
3 kDa MWCO
Legal Information
Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Alessandra Drusco et al.
EBioMedicine, 30, 105-112 (2018-04-13)
The World Health Organization has recently introduced molecular prognostic-diagnostic biomarkers in the classification of Central Nervous System (CNS) tumors. In order to characterize subclasses of tumors that cannot find a precise location in the current classification, and, or cannot be