Key Documents
Z740167
Millipore
Amicon® Ultra-2
Ultracel-100 membrane, 100 kDa
About This Item
UNSPSC Code:
40161500
material
regenerated cellulose membrane
styrene-butadiene housing
sterility
non-sterile
feature
100 kDa
packaging
pack of 24 ea
manufacturer/tradename
Millipore UFC210024
tube size
~2 mL
working volume
2 mL
pore size
100 kDa MWCO
Legal Information
Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)