Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z740149

Xtractite sorbent pellets for removal of heavy metals, radionuclides and actinides

pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113034
NACRES:
NB.35

packaging

pkg of 1 ea

manufacturer/tradename

Xplosafe, LLC 8001

General description

Xtractite sorbent pellets provide selective, rapid and economical removal of radionuclides, heavy metals and actinides. Xtractite sorbent contains a high density reagent on large surface area pellets and have a high affinity for metal adsorption of large quantities of heavy metal ions.

The high density reagent reduces radiation release from the radionuclide-loaded pellets while the refractory nature of the sorbent prevents radiation or heat damage to the loaded pellets.

Applications:
  • Water treatment
  • Treatment of radioactive effluents
  • Industrial waste water treatment
  • Environmental remediation
  • Analytical and research applications

Benefits:

  • Higher capacity for removal of heavy metals compared to polymer based resins
  • Little or no interference by lighter ions
  • Lower costs /volume compared to polymer based resins
  • Significantly more resistant to radiation and heat

Legal Information

Xtractite is a trademark of XploSafe, LLC

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service