Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z699160

Heterotrophic Plate Count (HPC) Broth with TTC

pack of 50 × 2 mL

Synonym(s):

broth media, hpc broth, media broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40151500
NACRES:
NB.79

packaging

pack of 50 × 2 mL

manufacturer/tradename

Maine Mfg 10496151

application(s)

food and beverages

General description

HPC is used to determine total count at incubation temperatures of 35°C. All bacteria develop on HPC with indicator media and produce a red color as a result of the precipitation of formazan following the reduction of 2,3,5- triphenyltetrazolium chloride (TTC) by bacteria.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service