Key Documents
Z699142
Whatman® Cetrimide Broth
Synonym(s):
broth media, difco agar, media broth
About This Item
UNSPSC Code:
41106225
packaging
pkg of 50 ea
manufacturer/tradename
Maine Mfg 10496146
General description
Pseudomonas aeruginosa is characterized by the production of pyocyanin (a blue green, water soluble, non-fluorescent, phenazine pigment) which is stimulated by the inclusion of magnesium chloride and potassium sulfate in the broth. Cetrimide (N-cetyl-NNN-trimethylammonium bromide) is added to inhibit bacteria other than Pseudomonas aeruginosa. Its action as a quaternary ammonium cationic detergent causes nitrogen and phosphorous to be released from bacterial cells other than Pseudomonas aeruginosa.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)