Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z699136

M-FC media

pack of 50 × 2 mL

Synonym(s):

fc agar, m-fc agar, mfc broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NB.79

packaging

pack of 50 × 2 mL

manufacturer/tradename

Maine Mfg 10496124

application(s)

food and beverages

General description

M-FC Broth allows the development of fecal coliforms at elevated temperatures (44.5°C).

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service