Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z699055

MRS Media in Ampoules

pack of 50 × 2 mL

Synonym(s):

bacteria growth media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111700
NACRES:
NB.79

packaging

pack of 50 × 2 mL

manufacturer/tradename

Maine Mfg 10496112

application(s)

food and beverages

General description

MRS medium supports luxuriant growth of all lactobacilli, even the slow growing species.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service