Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z699047

Wallerstein Broth

pack of 50 × 2 mL

Synonym(s):

broth media, media broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111700
NACRES:
NB.79

packaging

pack of 50 × 2 mL

manufacturer/tradename

Maine Mfg 10496108

application(s)

food and beverages

storage temp.

2-8°C

General description

WL Nutrient Broth is for the cultivation and enumeration of yeast and WL Differential Broth is for determination of bacterial count. Use of the medium at pH 5.5 and incubation at 25°C will give reliable counts for brewer′s yeast. Adjustment of the pH to 6.5 and incubation at 30°C allows for the selective growth of baker′s and distiller′s yeast.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service