Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z699012

M-Endo Coliform Broth

pkg of 50 ea

Synonym(s):

coli form, coliform test, coliforms test

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111700
NACRES:
NB.79

packaging

ampule of 50 × 2 mL
pkg of 50 ea

manufacturer/tradename

Maine Mfg 10496103 (GVS Life Sciences)

application(s)

food and beverages

storage temp.

2-8°C

General description

M-Endo is a red colored media, which needs to be stored in the dark to prevent discoloration. Gram-positive bacteria are inhibited on this media by the desoxycholate and lauryl sulfate. The addition of ethanol increases the antibacterial nature of the formulation. Lactose fermenting organisms form aldehydes, which react with Schiff′s reagent (basic fuchsin and sodium sulfite) to give red colored zones around the colonies. Coliform colonies are therefore red with a characteristic metallic sheen.

Packaging

Packaged in 2 mL ampules, 50 per box.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

pictograms

FlameHealth hazard

signalword

Danger

hcodes

flash_point_f

84.2 °F

flash_point_c

29 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service