Key Documents
Z698997
Whatman® M-Green Yeast and Mold Broth
Synonym(s):
agar broth, bacteria test, test bacteria
About This Item
UNSPSC Code:
41106227
Pricing and availability is not currently available.
packaging
pkg of 50 ea
manufacturer/tradename
Maine Mfg 10496101
General description
M-Green Yeast and Mold Broth is an improved modification of the liquid media. The addition of bromocresol green, which diffuses into fungal colonies as an alkaline reaction, allows them to be easily identified. Metabolic by-products from the developing colonies diffuse into the surrounding medium, further reducing the pH which aids in the inhibition of bacterial growth, but also produces an acid reaction that causes residual bromocresol green to change to yellow.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)