Z698083
Whatman® Nytran™ SuPerCharge (SPC) nylon blotting membranes
diam. 30 cm × 3 m, pore size 0.45 μm, pkg of 1 roll
Synonym(s):
blotting membrane, nylon membranes
About This Item
material
Nylon membrane (Nytran SuPerCharge (SPC))
packaging
pkg of 1 roll
manufacturer/tradename
Whatman 10416296
Whatman Article No. 28415380 (US reference)
diam.
30 cm × 3 m
pore size
0.45 μm
0.45 μm
suitability
suitable for (Southern and Northern blotting; colony and plaque lifts and Dot-/Slot-Blots)
General description
Nytran SPC membrane is cast uniformly on both sides of a support matrix, demonstrating excellent symmetry. This gives the membrane the ability to lie flat without curling. With typical manufacturing techniques, increasing positive charge tends to increase background. Nytran SPC membranes are manufactured using a process that allows the combination of high positive charge with low background. Whether using radioactive or nonradioactive detection techniques, Nytran SPC consistently gives high signal with extremely low background. Whatman Nytran nylon membranes are available in two formats: Nytran N is moderately charged and Nytran SuPerCharge (SPC) has a very high positive charge.
Other Notes
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
Legal Information
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)