Key Documents
Z694800
Whatman® Elutip-d DNA purification minicolumns
ELUTIP-D column
About This Item
UNSPSC Code:
41105500
Pricing and availability is not currently available.
packaging
pkg of 1 ea
manufacturer/tradename
Whatman 10462617
General description
The Elutip-d minicolumns are designed for purification of DNA with high recovery rates. They provide a simple and convenient method for purification of DNA in the15 bp to 50 Kb range.
The Elutip-d columns are ideal for removal of unincorporated nucleotides and other contaminants from radiolabeling reactions to reduce background levels and increase sample activity. They also provide an excellent method for isolation of nucleic acids from low-melt agarose gels.
The Elutip-d column matrix binds nucleic acids in high quantities upon sample application under low salt conditions. Contaminants are washed through the column and the purified DNA is then eluted with high salt. The eluted sample is ready for use in a variety of assays that require high purity of the nucleic acid.
Elutip-d columns are used with standard syringes. Optional prefilters contain non-binding cellulose acetate membranes and are designed to increase efficiency by removing gel pieces that could otherwise clog the column.
Features and Benefits
  • High recovery of single- and double-stranded DNA
  • Eliminates contaminants that can cause high background or interfere with sample activity
  • Sample is eluted in a small volume
  • 100 μg capacity
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)