Z694207
Troemner Lab-Frame standard kit with fiberglass rods
large, 48 in. x 48 in.
Synonym(s):
laboratory clamps, laboratory support rods
About This Item
size
L
manufacturer/tradename
Troemner 916195
base size
18 in.
frame L × W
48 in. × 48 in.
Looking for similar products? Visit Product Comparison Guide
General description
Components:
- 2 in. (51 mm) rods (qty 10)
- 18 in. (457 mm) rods (qty 3)
- 36 in. (914 mm) rods (qty 2)
- 48 in. (1219 mm) rods (qty 10)
- S-connectors (qty 38)
- Rod end connectors (qty 4)
- Lab-frame feet (qty 6)
- traditional center-less ground aluminum
- non-corroding fiberglass
- tough stainless steel
A range of lab-frames specifically designed to fit within laboratory fume hood include stainless steel rods. Nickel-plated connectors and lab-frame feet are standard in all kits. Lab-frame bases may be permanently mounted to bench top with screws (included).
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service