Z692972
Interlocking Puzzle Rack for 1.5/2.0, 15 and 50 mL tubes
L × W × H 151 mm × 135 mm × 66 mm, pkg of 4 ea
Synonym(s):
Tube rack, laboratory racks, test tube racks
About This Item
Recommended Products
feature
Holds 1.5, 2.0, 15 and 50 mL tubes
packaging
pkg of 4 ea
manufacturer/tradename
Heathrow Scientific HS29060A
L × W × H
151 mm × 135 mm × 66 mm
General description
Two-sided puzzle-shaped pieces hold an assortment of 1.5/2.0, 15 and 50 mL test tubes. Pieces can be used separately for small sample applications or connect in a square or row configurations. Imprinted references aid with easy retrieval of tubes.
- Bold assorted colors: yellow, blue, green and orange for quick identification
- Constructed of robust polypropylene
- Autoclavable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service