Key Documents
Z692433
Rachet tool, for 30 x 30 cm and 40 x 40 cm support jacks
pkg of 1 ea
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
packaging
pkg of 1 ea
General description
An optional racheting tool is available to add extra leverage to the actuating rod on 12 x 12 in. and 16 x 16 in. jacks for easy, accurate adjustments.
Choose from one of the most recent versions:
Certificates of Analysis (COA)