Z691585
Mini Bin™ aluminum foil dispenser
L × W × H 19 mm × 10 mm × 12 mm
Synonym(s):
aluminum foil, tube foil, tube sealing foil
About This Item
manufacturer/tradename
Heathrow Scientific HD23520A
L × W × H
19 mm × 10 mm × 12 mm
General description
For use with 102 mm or 153 mm (4 or 6 inch) foil rolls (sold separately).
- Safe - unique cutter guard with safety blade
- Compact - for limited spaced work areas
- Stable - non-slip base
- Versatile - holds 102 mm or 153 mm (4 or 6 inch) foil rolls
- Unique - foam strip prevents foil from retracting back into the unit
- Available separately - Z691569, 102 mm (4-inch) Foil Roll and Z691577 153 mm (6-inch) Foil Roll
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service