Key Documents
Z691054
Clipox ground glass joint clip for conical joints
size NS10
Synonym(s):
clamp clips, glass clips, joint clips, steel clamps
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
material
stainless steel
size
NS10
manufacturer/tradename
Clipox 10010
General description
Clipox are safe, universal clips for ground joints manufactured from high grade stainless steel. Their slim, symmetric design solves application problems of common clips and clamps. They are more secure, have better thermal and chemical resistance and are able to be used with stand clamps.
Clipox are available to secure conical, spherical, stopcocks and glass stopper ground joints.
  • Increases the safety of glass instruments
  • Secure horizontal and vertical in case of pressure overload
  • Easy to mount and to remove due to two-sided handles
  • Longevity of suspension and shape due to spring steel wire
  • Outstanding thermal, chemical and acid resistance
  • Reduced scratch of glass surface due to polished surface of material
  • Made in Germany, production certified according to ISO 9001
Legal Information
Clipox is a trademark of Clipox Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)