Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z691003

Whatman® cellulose chromatography papers

2668 Chr sheets, 58 x 60 cm, 100/pk

Synonym(s):

cellulose chromatography papers, chromatography papers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900

material

cellulose membrane
sheets

grade

for chromatography

sterility

non-sterile

packaging

pack of 100

manufacturer/tradename

Whatman 10382461
Whatman Article No. 28414857 (US reference)

parameter

400 s / 100 mL flow rate

W × L

58 cm × 60 cm

basis weight

700 g/m2

suitability

suitable for chromatography

Looking for similar products? Visit Product Comparison Guide

Other Notes

Global SIAL customers (excluding Japan) can now order any product from the Cytiva Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.

The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susana Campuzano et al.
Applied and environmental microbiology, 75(18), 5972-5980 (2009-07-28)
A Streptococcus mitis genomic DNA fragment carrying the SMT1224 gene encoding a putative beta-galactosidase was identified, cloned, and expressed in Escherichia coli. This gene encodes a protein 2,411 amino acids long with a predicted molecular mass of 268 kDa. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service