Z690716
Whatman® glass microfiber filters, Grade 934-AH®
diam. 24 mm
Synonym(s):
glass fiber filter papers, glass microfiber filters
About This Item
material
borosilicate glass membrane
circles
description
basis weight, 64 g/m2
manufacturer/tradename
Whatman 1827-024
Whatman Article No. 28418465 (US reference)
parameter
47 speed (filtration)
diam.
24 mm
size
thickness
435 μm
pore size
1.5 μm retention
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Water pollution monitoring
- Air pollution monitoring
- Cell harvesting
- Liquid scintillation counting
Other Notes
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
Legal Information
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service