Z690651
BactiZapper™ infrared microsterilizer
AC/DC input 230 V AC, UK plug (3 flat pins)
All Photos(1)
About This Item
UNSPSC Code:
41103200
Recommended Products
feature
CE compliant
manufacturer/tradename
Benchmark Scientific B1000-UK
General description
The BactiZapper has an asbestos-free core element that uses infrared heating to produce 815°C (1500°F).
It completely sterilizes microorganisms in 5-7 seconds. The BactiZapper uses well-proven technology to provide instant sterilization of platinum inoculating loops, needles, glass tube/pipette mouths and various metal & borosilicate glass instruments, commonly used in microbiology research.
The ceramic heating element is safely enclosed in a perforated stainless steel guard chamber, protecting users from accidental contact with the extreme temperatures of the core area. It is extremely compact, convenient and can be used in anaerobic and aerobic chambers.
It completely sterilizes microorganisms in 5-7 seconds. The BactiZapper uses well-proven technology to provide instant sterilization of platinum inoculating loops, needles, glass tube/pipette mouths and various metal & borosilicate glass instruments, commonly used in microbiology research.
The ceramic heating element is safely enclosed in a perforated stainless steel guard chamber, protecting users from accidental contact with the extreme temperatures of the core area. It is extremely compact, convenient and can be used in anaerobic and aerobic chambers.
Legal Information
BactiZapper is a trademark of Benchmark Scientific, Inc.
accessory
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)