Key Documents
Z690643
BactiZapper infrared microsterilizer
AC/DC input 115 V AC
About This Item
UNSPSC Code:
41100000
Product Z690643 is not currently sold in your country. Contact Technical Service
feature
CSA approved
manufacturer/tradename
Benchmark Scientific B1000
General description
The BactiZapper has an asbestos-free core element that uses infrared heating to produce 815°C (1500°F).
It completely sterilizes microorganisms in 5-7 seconds. The BactiZapper uses well-proven technology to provide instant sterilization of platinum inoculating loops, needles, glass tube/pipette mouths and various metal & borosilicate glass instruments, commonly used in microbiology research.
The ceramic heating element is safely enclosed in a perforated stainless steel guard chamber, protecting users from accidental contact with the extreme temperatures of the core area. It is extremely compact, convenient and can be used in anaerobic and aerobic chambers.
Legal Information
BactiZapper is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)