Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z690236

Welch® silicone tubing cartridge kit for bio-fluids

diam. 1.6 mm, flow rate 0.003-0.03 L/min

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105108
NACRES:
NB.79

manufacturer/tradename

Welch 3432K-01

parameter

0.003-0.03 L/min flow rate

diam.

1.6 mm

Looking for similar products? Visit Product Comparison Guide

General description

Packs of 5 ready assembled tubing cartridges that clip directly into pumping head using Welch Snap-grip clamps.

Legal Information

Welch is a registered trademark of Gardner Denver Thomas, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service