Z690201
Welch® peristaltic pump
digital, AC/DC input 115 V AC
About This Item
feature
CSA approved
manufacturer/tradename
Welch 3200B-01
parameter
0.003-0.3 L/min flow rate
20-200 rpm speed
Looking for similar products? Visit Product Comparison Guide
General description
- Complete system - pump head, tubing & drive
- Lok-n-Flow tubing cartridges
- Bi-directional flow control & max flow button
- Variable speed
- Stackable & portable
Starter Kit 3200K-01 is included with every pump
Initial Tubing Cartridges:
- 1.6 mm silicone (2 each)
- 3.2 mm silicone (2 each)
- 4.8 mm silicone (2 each)
- 4.8 mm PVC Tubing (25 feet)
- Hose Clamps (4 each)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service