Z685496
Vacuubrand chemistry pumping units, automatic control, Model PC 610 NT
AC/DC input 115 V AC, US 2-pin plug
All Photos(1)
About This Item
UNSPSC Code:
41105101
NACRES:
NB.79
manufacturer/tradename
Vacuubrand 737103
parameter
1.5 mbar vacuum
3.4-3.8 m3/h pumping speed
General description
Chemistry pumping units of this series are well proven for vacuum generation and control for many evaporation processes. The popular two-stage MZ 2C NT chemistry diaphragm pump is the heart of these pumping units, frequently used for medium sized vacuum applications involving "common" solvents.
The pumping units are equipped with a CVC 3000 vacuum controller with a solenoid valve for electronic vacuum control. The exhaust waste vapor condenser is outstandingly efficient and compact. It provided near 100 percent recovery of solvents, for economical recycling and environmental protection.
The pumping units are equipped with a CVC 3000 vacuum controller with a solenoid valve for electronic vacuum control. The exhaust waste vapor condenser is outstandingly efficient and compact. It provided near 100 percent recovery of solvents, for economical recycling and environmental protection.
- Optimized vacuum even with gas ballast
- Easily operated controller with clear text menus with integrated venting valve
- Chemistry design flow control valve with large cross section for unrestricted performance
- Whisper quiet and very low vibration
- Excellent environmental friendliness due to efficient solvent recovery
Choose from one of the most recent versions:
Certificates of Analysis (COA)