Key Documents
Z685151
Sigma-Aldrich
Crucible Cradle
with 3 ceramic triangles to fit crucibles 5-10 mL, 15-25 mL, or 30-40 mL
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
General description
Eliminates the need for crucible tongs. Users can safely heat, cool, and weigh crucibles without having to touch the product. Unique locking slots in the cradle keep ceramic triangles and wire gauze and other accessories safely in place. No more accidental snagging which leads to crucible breakage.
Features:
  • Three threaded steel legs to provide support during use
  • Removable wooden handle protects fingers when heating
  • Includes three locking ceramic triangles to fit crucibles 5 to 40 mL
  • Optional locking wire gauze, evaporating dish holder, replacement triangles, and micro crucible holder
Legal Information
Crucible Cradle is a trademark of Global Market Connections, Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)