Key Documents
Z684732
Ace spherical reaction flask with three vertical necks
capacity 1 L
Select a Size
1 EA
$464.00
$464.00
Check Cart for Availability
Select a Size
Change View
1 EA
$464.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$464.00
Check Cart for Availability
material
borosilicate glass
spherical
three-neck (Vertical)
three-neck
joint
Joints ST/NS24/40 (3)
description
0-10 mm zero dead space bottom valve
sterility
non-sterile
feature
cap: no
packaging
case of 1
manufacturer/tradename
Ace Glass 6483-110
capacity
1 L
General description
Heavy wall, spherical reaction flask with three vertical necks. Flask has a Zero Dead Space (ZDS) bottom valve. The valve features no material hang-up in the fully open position. When closed, the valve seats flush with the bottom of the flask.
Choose from one of the most recent versions:
Certificates of Analysis (COA)